GeneMapper® ID-X Getting Started
Implementing GeneMapper® ID-X in your lab is easy because GeneMapper® ID-X is built on the same trusted platform as the GeneMapper® ID-X product suite. Forensic DNA analysts will find the interface intuitive and familiar; and, most importantly, the key peak detection and sizing algorithms remain unchanged. This enables laboratories that have already validated the current version of GeneMapper® ID-X to easily implement the new software. In addition, a comprehensive help system Tutorial Reference Guide is built into the software to guide the users through set up and operation and Applied Biosystems will offer online webinars and hands-on course tutorials. Implementation and training resources include:
- Getting Started Guide tutorial that demonstrates key functionalities of GeneMapper® ID-X
- Reference Guide that provides algorithm and quality value details
- Quick Reference Guide
- Online Context Specific Help built within the software
- Free introductory online webinars
- In-depth 3-4 day hands-on course
- Intel® Core™ i3-3240 CPU @ 3.40GHz or newer
- 8 GB RAM
- 10 GB available on boot drive
- DVD-ROM drive
- 10/100 NIC with RWU (internal) or above installed
- TCP/IP installed
Windows® 7 Professional, 64-bit (Service Pack 1)