Invitrogen eBioscience Super Bright 436 dye is a polymer-based fluorophore excited by the violet laser (405 nm) that emits signal at 436 nm with high efficacy. The technology behind the Super Bright polymer dye family makes them useful for identifying low-abundance markers on a cell surface in flow cytometry.
An alternative to Brilliant Violet 421 conjugates, antibody conjugates of Super Bright 436 provide increased resolution of positive and negative populations, but with less background and less interfering dye–dye interaction than Brilliant Violet 421 conjugates. In addition, antibody conjugates of this polymer dye are significantly brighter than Invitrogen eFluor 450 conjugates and Pacific Blue conjugates.
Executing a well-designed experiment requires multiple reagents. Super Bright conjugates are fully compatible with Invitrogen UltraComp eBeads microspheres for compensation, viability stains, fluorescent dyes and other reagents commonly used in flow cytometry. Robust, highly sensitive, and stable, Super Bright conjugates easily fit into any flow cytometry panel.
We offer Super Bright 436 dye conjugated to primary antibodies for use in flow cytometry. 
Super Bright 436 dye dashboard
Initial brightness
The violet laser–excitable Super Bright 436 dye is optimized for use in flow cytometry applications, but can also be used for high-abundance targets in fluorescence microscopy
Photostability in buffer
  405 450/50 414 436  
Laser line
Common filter set
Excitation max
Emission max
Super Bright 436 dye products
We offer Super Bright 436 dye conjugated to primary antibodies for use in flow cytometry.
Learn more here:
Primary antibodies for the violet laser
Invitrogen eBioscience Super Bright polymer dyes are violet laser–excitable fluorophores that expand the capabilities of multi-parametric flow cytometry analysis. This family of bright fluorophores enables the use of more markers off the violet channel and increases the detection of low-abundance antigens due to their super bright signals.
The Super Bright polymer dyes join the trusted Invitrogen Alexa Fluor dye and Invitrogen eFluor dye portfolios in providing fluorochromes compatible with the 405 nm spectral line of the violet laser.
Resources
Primary antibodies for the violet laser
Stain your own cell by using our simulation tool to develop reproducible results with many of our signature fluorescent dyes. Develop your multiplexing strategy for antibody probes, reactive dyes, and counterstains to create your perfectly labeled fluorescent cell.
Learn more about primary antibodies for the violet laser
Fluorescence SpectraViewer
Plot and compare spectra and check the spectral compatibility for common fluorophores including all of the fluorescent probes from Thermo Fisher Scientific. The Fluorescence SpectraViewer tool is formatted for both microscope imaging and flow cytometry applications.
Go to the Fluorescence SpectraViewer