Bioprocessing Fluid Management
Streamlining fluid management across bioprocesses
Help ensure high performance and reliability in various stages of your development process with Thermo Scientific fluid management solutions. From bioprocess bags, bottles, and carboys to filters and tubing assemblies, secured at connection points with the BioTitan Retention Device, our customizable products are designed to safeguard workflows against leaks, contamination, and supply disruptions.
Our global single-use manufacturing network offers in-region and assembly manufacturing capabilities designed to strengthen supply assurance. In addition to dedicated technical support for your single-use fluid management needs, we offer the Single-Use Assembly Configurator to help simplify both standard and custom assembly designs. This intuitive digital tool, which leverages our Global Standard Component Library, enables you to create bioprocess bag and tubing assemblies with greater design efficiency and faster delivery times.
Fluid management solutions to empower results
Bioprocess bags
Ready-to-use bioprocess bags designed for sterile liquid handling, available in scalable chamber formats made from high-performance films like Aegis5-14 and CX5-14 and supported by multiple port design options.
Bioprocess bottles and carboy
Low and ultra-low particulate bottles and carboys support optimal storage and transport of biological and chemical solutions. Available in multiple sizes and made from high-quality materials, they also offer standard or custom assembly systems.
Fluid transfer solutions
Reduce leaks and contamination risk with sterile tubing and adaptors secured by the BioTitan Retention Device. Standard fluid transfer assemblies streamline setup and drive time and cost efficiency throughout your workflow.
Production tanks and drums
Trusted, robust solutions for large-volume storage and transport to accommodate a variety of sizes and processes. Single-use tank liners are designed specifically to fit Nalgene and Thermo Scientific cylindrical tanks and drums so you can enhance your mixing capabilities and maximize the utility of your carboys and tanks.
BioTitan Retention Device
The BioTitan Retention Device enhances single-use assemblies with a secure 360° seal around tubing and fittings. As a novel replacement for traditional cable ties, it helps mitigate the risk of leaks, simplify setup, and improve the reliability and integrity of bioprocessing workflows.
Fill finish solutions
Standard and configurable fill-finish solutions to help maximize drug product recovery.  Exceptional lead times due to standard and configurable product design.
Bioprocessing solutions designed for end-to-end productivity
Bioreactors
Engineered for optimal cell culture and fermentation processes, enabling precise control over environmental conditions. Bench scale, rocker, and single-use bioreactors support your project from process development to commercial manufacturing.
Cell factories
Easily scale your adherent cell cultures from flasks to Nunc High Density Cell Factory Systems. Available from single layers to 52 layers of surface to support expansion. Filter assemblies, tubing, special caps, and connectors are available to help create a closed system.
Single-use bioprocessing equipment
Efficiently grow and harvest your cells with equipment specially designed for your application like DynaSpin Single-Use Centrifuge for bioprocessing. It features a unique single use rotor and can replace primary filtration and reduce burden on secondary filtration.
Single-Use Configurator Tool
Build your own 2D, 3D bioprocess bags, and fluid transfer assembly solutions with our Single-Use Assembly Configurator, an easy-to-use online design tool. With its comprehensive selections, real-time 3D visualization, and promptly issued quotes, you can streamline your design process to save valuable time today.
Frequently asked questions
Bioprocessing fluid management involves transferring bioprocess fluids such as media, buffers, and samples within bioprocessing facilities using tubing, filters, bags, bottles, and carboys. Preventing leaks and contamination is crucial, as they can result in costly batch losses and facility contamination. To address this, our products undergo rigorous integrity and leak analysis testing. The novel Thermo Scientific BioTitan Retention Device secures fittings and connections with 360° seal points, significantly reducing the risk of leaks.
Single-use systems generally include a frame, mechanical components, and disposable elements. The parts of the system that contact samples are disposable bags, containers or tubing, typically made of plastic. These pre-sterilized bags and fluid transfer assemblies can be easily replaced after each use, to help minimize contamination risk and help reduce time on cleaning and decontaminating the equipment. Our single-use bags are made from the single-use Thermo Scientific Aegis5-14 film, which is Irgafos-free to support healthier cell cultures.
Facilities face many challenges from finding the correct solutions to quality and supply assurance. To overcome these challenges, we offer a Single-Use Assembly Configurator and specialist support to help you design your ideal assembly. Our products are rigorously tested for integrity and made from quality materials. Rather than using cable ties with uneven compression at connection points that can result in leaks, secure your single-use assemblies with the BioTitan Retention Device that forms a 360-degree seal, mitigating the risk of connection failures. To help ensure supply, we have a large single-use global manufacturing network to offer an in stock and ready-to-ship portfolio to help reduce lead times.
We offer a comprehensive line of products designed to work together to help you scale from 5 mL to 500 L. Our bioprocess bags are available in a wide variety of shapes and sizes to support many applications across the workflow from fluid mixing to fill finish. Many of our products are stocked and ready to ship. We have a Global Standard Component Library (GSCL) that can be leveraged to support your custom needs. Find solutions for your application using the Single-Use Assembly Configurator or by contacting a specialist.
Single-use technologies can support sustainability goals by offering a means to significantly reduce the amount of cleaning fluids, water, and energy used in production. We have also introduced biobased products and reduced packaging to lower our combined environmental footprint. To further reduce your environmental impact, you can speak with one of our bioprocessing specialists who can help you optimize your processes for efficiency and sustainability. Keep track of your progress by enrolling in our Bioproduction Sustain Program. As part of this program, you will receive a detailed annual report that quantifies the emissions reduction achieved from your eligible product purchases to help you achieve your sustainability goals.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.