Neurobiology Antibodies
We have a diverse array of highly specific antibodies for neuroscience research that have been validated* for multiple applications, including protein targets important in the study of glia, protein trafficking, neurogenesis, axon guidance, dendrite development, neuronal migration, growth factors, neuromuscular junction, neurotrophins, apoptosis, and synaptogenesis.
Download the Invitrogen Neurobiology Antibodies poster
Already know your neurobiology research target of interest?
Find antibodies of interest using the search tool below. Then filter the results by target or host species, monoclonal or polyclonal antibody type, and other criteria.
Search for antibodies
Primary Antibodies reference component
Tau antibody immunohistochemistry image
Neurodegenerative disease research targets
Neurodegenerative research targets include cleavage specific antibodies to amyloid beta, phosphospecific, and pan antibodies against tau, parkin, LRRK2, nuclear aggregation of huntingtin (HTT) and MBP. Targeted neurodegenerative diseases include: Alzheimer’s disease, amyotrophic lateral sclerosis, Huntington’s disease, Multiple sclerosis, and Parkinson’s disease.
Markers of interest: beta amyloid, C9orf72, HTT, IL-17A, alpha synuclein
Synaptophysin antibody immunofluorescence image
Neuron markers
Neuron markers allow the specific identification of different neuronal cells in samples. Specific neuronal subtypes, like dopaminergic, glutamatergic and motor neurons express specific proteins that allow them to be specifically detected in imaging and flow cytometry experiments.
Markers of interest: ChAT, OTX2, GAD65, NMDAR2B, ISL2, calretinin
A2B5 antibody immunofluorescence image
Glial progenitor cell markers
Glial progenitor cells can be differentiated from neural stem cells, and then further differentiated into astrocytes and oliogodendrocytes. These progenitor cells are characterized by expression of CD44 and A2B5.
Markers of interest: A2B5, CD44
MAP2 antibody immunofluorescence image
Presynaptic and postsynaptic markers
Neuron communication involves the release of neurotransmitters from the presynaptic cells to the post synaptic cells. These cells express specific markers like synaptophysin (presynaptic) and PSD95 (postsynaptic) that allow for the differentiation of cell types.
Markers of interest: HOMER1, PSD95, synaptophysin, synaptotagmin, VGLUT1
GalC antibody immunofluorescence image
Oligodendrocyte progenitor cell and oligodendrocyte markers
Oligodendrocyte cells support and insulate neurons, creating the myelin sheath. They can be differentiated from glial progenitor cells. Important markers like GalC can be used to identify oligodendrocytes in a variety of applications and sample types.
Markers of interest: NG2, OLIG1, OLIG2, PDGFRA, MBP
Germlayer staining image
Neuroectoderm markers
Mammalian neurogenesis begins with the induction of neuroectoderm, which forms a neural plate and then folds to give rise to the neural tube. These structures are made up of a layer of neuroepithelial progenitors (NEPs).NEPs can be rapidly turned into primitive neural stem cells (NSCs).
Markers of interest: NCAM, SOX2, OTX2, PAX3, PAX6
Germlayer staining image
Neural stem cell markers
Neuronal stem cells are the first step in neuronal development from embryonic stem cells or induced pluripotent stem cells. The express important markers, nestin and SOX1 in addition to others. Neuronal stem cells can be further differentiated into glial progenitor cells and neuronal progenitor cells.
Markers of interest: SOX1, SOX2, PAX3, PAX6, FOXG1
Astrocytes stained cell image
Astrocyte markers
Astrocytes cells support the neural network. They express neurotransmitter receptors and may possibly play a role in neuronal communication. They express the important marker GFAP and can be differentiated from glial progenitor cells.
Markers of interest: ALDH1L1, EAAT2, GFAP, glutamine synthetase, S100beta
IBA1 antibody immunohistochemistry image
Microglia markers
Microglia play a major role in the immune system in the central nervous system, phagocytosing foreign material and cellular debris. Microglia can activate pro-inflammatory cytokines like, IFN-gamma, IL1-alpha, and TNF-alpha. Microglia also synthesize amyloid precursor protein (APP) in response to injury.
Markers of interest: CD11b, CD68, CX3CR1, HLA-DR, IBA1
*The use or any variation of the word “validation” refers only to research use antibodies that were subject to functional testing to confirm that the antibody can be used with the research techniques indicated. It does not ensure that the product(s) was validated for clinical or diagnostic uses.
We offer a diverse collection of Invitrogen products for imaging and dissecting neural networks and their functions, including neuronal tracers and neural anatomy and function probes. These dyes and stains are available in array fluorescent colors,giving you the flexibility to design your fluorescence imaging experiments.
Our Invitrogen products offer a broad technology platform for studying neural stem cells (NSC), providing you with innovative solutions that help enhance basic research and guide therapeutic discovery. These include neural stem cell antibodies, cell lines, media and reagents as well as neural stem cell growth factors and cytokines.