antibodies-animal-showcase
Our anti-hamster secondary antibodies are affinity-purified polyclonal antibodies with well-characterized specificity for hamster IgG. They are available in host species including goat and rabbit, and in the following formats:
  • Unlabeled antibodies
  • Enzyme conjugates (alkaline phosphatase (AP) or horseradish peroxidase (HRP))
  • Fluorescent conjugates (Alexa Fluor, DyLight, Fluorescein (FITC), and more)
  • Biotin
View all anti-hamster secondaries
Our secondary antibodies are available as whole IgG or the F(ab')2 fragment.  Each of our antibodies is affinity purified using its target antigen coupled to an agarose support. This affinity purification eliminates nonspecific antibodies, resulting in low background signal. Conjugated antibodies are also affinity purified before labeling.
Selected secondary antibodies have been further purified by adsorption of the purified antibody to an affinity column containing immobilized serum proteins or the IgG of selected species. This additional processing step minimizes antibody cross-reactivity to the serum proteins of those selected species. These antibodies are designated as cross adsorbed or highly cross adsorbed, based on the amount of processing which has occurred.
How to find a secondary antibody
Step 1: Use the tool at the top of the page to filter by target, application, target species
Step 2: View more filters to narrow results by host species, conjugate and other criteria
See all secondary antibodies
bulk-pod-showcase-270x195
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.  
Complete request form