Biotin Affinity Purification
We offer a variety of resins for the purification of biotinylated or desthiobiotinylated proteins, peptides, and other molecules. These resins are available in multiple pack sizes, as well as in spin columns, kits, and coated plates for some ligands. Select products based on avidin, streptavidin, NeutrAvidin™, or CaptAvidin™ proteins.
- Product choice—a variety of biotin binding choices to meet your purification needs
- High performance—resins that maximize binding capacity and elution conditions
- Flexible—available in multiple pack sizes
Choose the right biotin-binding resin for your experiment
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
|
---|---|---|---|---|---|---|
Avidin Agarose | Streptavidin Agarose | Streptavidin Agarose HC | NeutrAvidin Agarose | NeutrAvidin Agarose HC | Monomeric Avidin Agarose | |
Biotin binding sites | 4 | 4 | 4 | 4 | 4 | 1 |
Binding capacity (biotinylated BSA) | ≥20 µg/mL biotin | 1–3 mg/mL | >10 mg/mL | 1–2 mg/mL | >8 mg/mL | >1.2 mg/mL |
Specificity | Low | Higher | Higher | Highest | Highest | High |
Nonspecific binding | High | Lower | Lower | Lowest | Lowest | Low |
Elution conditions | Harsh | Harsh | Harsh | Harsh | Harsh | Mild |
20219 (5 mL), 20225 (5 x 5 mL) | 20347 (2 mL), 20349 (5 mL), 20353 (10 mL) | 20357 (2 mL), 20359 (5 mL), 20361 (10 mL) | 29200 (5 mL), 29201 (10 mL) | 29202 (5 mL), 29204 (10 mL) | 20228 (5 mL), 20267 (10 mL) |
Need a coated microplate to capture biotinylated molecules for ELISA development and other plate-based assays?
We also offer a wide range of biotinylation reagents for proteins, peptides, antibodies, and other molecules.
For Research Use Only. Not for use in diagnostic procedures.