Search
| Label (Ex/Em) * | Cat. No. |
|---|---|
| Dansyl (336/517) | D57 |
| Marina Blue (365/460) | M12652 |
| Pacific Blue (410/455) | P22652 |
| NBD (463/536) | N360 |
| Fluorescein (496/519) | F362 |
| Oregon Green 488 (501/526) | O12650 |
| BODIPY FL (505/511) | D3800 |
| BODIPY 530/550 | D3815 |
| Tetramethylrhodamine (540/566) | T1391 |
| Lissamine rhodamine (560/581) | L1392 |
| Texas Red (582/601) | T1395MP |
| Biotin (<250/none) | B1550, B1616 |
| * Fluorescence excitation (Ex) and emission (Em) spectral maxima, in nm, are in methanol. The spectra may be different in membranes. | |
For Research Use Only. Not for use in diagnostic procedures.