Search
Immunofluorescence analysis of CBX3 in Hela Cells and CNE2Z Cells using Anti-CBX3 Recombinant Mouse Monoclonal Antibody (Cat. no. 730045). Hela Cells and CNE2Z Cells were fixed with 3% Formaldehyde and lysed with 1% TritonX-100. Cells were then labeled with Anti-CBX3 Recombinant Mouse Monoclonal Antibody (Cat. no. 730045) and detected with Goat-anti-human Alexa Fluor dye F488 (green). FLAG Cells were double labeled with Anti-CBX3 Recombinant Mouse Monoclonal Antibody (Cat. no. 730045) and detected with Goat-anti-human Alexa Fluor dye F488 (green) as well as with Mouse-anti-FLAG-M2 and detected with Goat-anti-Mouse Alexa Fluor dye F647 (yellow). The DNA in the nucleus was detected with DAPI (Blue). Control means cells were labeled only with secondary antibody Alexa Fluor Dye.