The genus Salmonella belongs to the family Enterobacteriaceae. Salmonella are Gram-negative, non-spore forming rods. There are over 2,500 serovars of Salmonella, which are characterized according to somatic (O) and flagellar (H) antigens.
Salmonella enterica is one of the most significant species responsible for human infection, and it can infect humans through various routes, including contaminated food and water.
A free guide to Salmonella test methods for food testing laboratories
Salmonellosis is the most frequently reported cause of foodborne illness. An estimated one million cases occur annually in the United States; of these, approximately 35,000 are laboratory-confirmed cases reported to the Centers for Disease Control and Prevention. Both animals and food products can become infected with Salmonella, leading to transmission to humans.
The ingestion of 10-100 cells can constitute a human infectious dose. Humans are highly susceptible to infection, and even low doses can infect and cause illness. Lower cell numbers can still be highly infectious, especially in high fat foods where the microorganism can escape the gastric acidity and be released in the intestine through bile-mediated dispersion of the lipids.
Applications
Despite the general perception that poultry and egg products are the primary source of Salmonella infections, many outbreaks in recent years have been associated with fresh produce, peanut butter, sprouted seeds, and fruits. Other affected foods include raw meat, powdered infant formula, milk and dairy products, fish, seafood, salad dressing, cake mixes and chocolate. Fruits, like other ready-to-eat foods, require routine Salmonella testing to ensure food safety.
Solutions
We offer a wide range of products and solutions for the detection, enumeration, and identification of Salmonella in foods and associated sample types.
FAQs
Salmonella is linked most often to raw or undercooked poultry, eggs, meat, and unpasteurized milk, but vegetables, spices, nuts, seafood, and other foods can also carry the bacteria. Many animals (including chickens, reptiles, and pets) can shed Salmonella that reaches food via faecal routes in water, fields, hands, and equipment—so food safety depends on strong hygiene and control across the supply chain.
The most common route is fecal contamination of food or water; this is why sanitation, clean irrigation, and hygienic slaughter are central to disease control and prevention programs worldwide. In low‑moisture or processed foods (for example, peanut butter or powdered ingredients), Salmonella may survive desiccation and remain present for extended periods.
Important: Even if contaminated food looks or smells normal, it can still cause salmonella infection (sometimes called food poisoning or salmonella poisoning). High‑risk groups for severe illness include young children, older adults, and people with a weakened immune system. Most people who contract Salmonella experience mild symptoms and recover without complications.
The microbiological testing “gold standard” is culture: pre‑enrichment (e.g., buffered peptone water), selective enrichment (e.g., Rappaport‑Vassiliadis, tetrathionate, or selenite cystine), plating to selective agars, and biochemical confirmation with identification to serovar as needed. Culture‑based methods typically require ~4–5 days for presumptives.
A widely recognized protocol was ISO 6579:2002 for detecting Salmonella in food and animal feed; today it’s superseded by ISO 6579‑1:2017 (Part 1: Detection of Salmonella spp.) which remains a key global reference across food samples and environmental swabs.
These culture methods are accepted by FDA (BAM) and USDA‑FSIS and remain foundational because they yield isolates for serology, antimicrobial testing, and regulatory control actions.
Yes. Validated rapid methods combine an enrichment step with molecular methods or immunoassays to speed Salmonella detection. Options include immunomagnetic separation, EIA/enzyme linked immunosorbent assay, ELISA with fluorescent or colorimetric detection, lateral flow immunoassays, and real‑time PCR. Depending on the matrix and enrichment design, test results can be available in ~24–48 hours (often after a 24 hour pre‑enrichment). Many workflows still confirm positives by culture.
In the food industry, although PCR is widely used for environmental surveillance (e.g., drains and equipment) to screen for Salmonella spp., it is proven to give accurate results for finished goods too. Rapid detection is accelerated by enrichment, but some food samples contain inhibitory compounds, so modified pre‑enrichment or matrix‑specific protocols may be needed to avoid false positives.
1) Sampling & pre‑enrichment. After compositing samples, labs pre‑enrich (commonly in buffered peptone water or modified variants) to resuscitate stressed cells.
2) Selective enrichment. Aliquots move to selective broths such as selenite cystine, tetrathionate, or Rappaport‑Vassiliadis to favor Salmonella over competing bacteria.
3) Plating & screening. Streak to XLD/HE/BS agars and examine suspect colonies.
4) Confirmation. Perform biochemical and serological tests and, if applicable, molecular confirmation. 
The organism thrives in animal‑origin food (raw poultry, eggs, meats) but can also contaminate produce and dry ingredients. You cannot see or smell Salmonella; only testing can determine if it’s present. Food processors typically integrate routine swabbing and lot testing to verify control.
For food and environmental monitoring, culture plus confirmatory testing is the regulatory benchmark because an isolate enables serotyping and epidemiology. For high throughput screening, validated molecular methods and immunoassays provide sensitive, specific detection when paired with proper enrichment, with culture used for confirmation.
Traditional culture needs multiple incubation steps, so presumptive identification often takes 4–5 days. With rapid methods (e.g., qPCR, LAMP, ELISA) run after 24 hour enrichment, screens can be completed by day 2; presumptive positives undergo culture confirmation. 
Yes. Environmental swabs and water samples are enriched and screened much like food. Many programs use PCR for rapid screening of drains, floors, and equipment as part of routine disease control and verification, with culture used for confirmation.
Both traditional and rapid testing workflows rely on pre‑enrichment and selective enrichment to amplify low numbers and resuscitate injured cells. This is critical because competing bacteria, low moisture, acids, or phenolics can suppress growth; matrices with inhibitory compounds may require modified pre‑enrichment to ensure reliable detection. 
Salmonella infection, a common cause of food poisoning, occurs when people consume food contaminated with Salmonella bacteria. These bacteria are often found in foods such as eggs, poultry, and vegetables, but can also be present in other foods if proper food safety measures are not followed. Symptoms of Salmonella infection typically appear within 6 to 72 hours after eating contaminated food and can include stomach cramps, abdominal pain, diarrhea, fever, and vomiting. While most cases of salmonella infections are mild and resolve on their own, some individuals—especially young children, older adults, and those with a weakened immune system—may experience more severe illness.
Diagnosis of Salmonella infection is usually based on the presence of symptoms and a physical exam by a healthcare provider, who will check for signs of dehydration and other symptoms such as severe diarrhoea or persistent abdominal pain. To confirm the infection, a stool sample is often collected and tested for the presence of Salmonella spp. using laboratory methods. Modern diagnostic techniques include enzyme linked immunosorbent assay (ELISA), molecular methods, and rapid detection tools such as colorimetric detection, which can quickly identify Salmonella bacteria in stool or other samples. In certain cases, especially if the infection is severe or has spread beyond the intestines, blood tests may be performed to detect Salmonella bacteria in the bloodstream.
If you suspect you have a Salmonella infection, it is important to seek medical attention, particularly if symptoms are severe or do not improve. A healthcare provider can recommend the best course of treatment, which may include oral rehydration solutions to replace lost body fluids and medications to manage symptoms. In rare cases, antibiotics or hospitalization may be necessary, especially for those at higher risk of complications.
Food processors and manufacturers play a vital role in preventing Salmonella contamination by implementing rigorous microbiological testing and food safety protocols. Regular testing of food samples for Salmonella enteritidis and other serovars helps ensure that processed foods are safe for consumption. Consumers can also help prevent Salmonella poisoning by practicing good hygiene, cooking foods thoroughly, and avoiding cross-contamination in the kitchen.
Disease control efforts, including public education about the risks of Salmonella infection and the importance of proper food handling, are essential for protecting human health. Drinking plenty of fluids, such as water or oral rehydration solutions, can help prevent dehydration during illness and support recovery. By working together—healthcare providers, food processors, and consumers—we can reduce the incidence of Salmonella infections and safeguard public health through effective prevention, rapid detection, and timely treatment.
For Research Use Only. Not for use in diagnostic procedures.