Search
Fluid transfer assemblies are critical components across the bioprocessing workflow. In the production of biologics, such as therapeutic proteins, fluid transfer facilitates various processes including cell culture media preparation, cell expansion, biologic harvest, purification, and fill-finish. Designed to manage large volumes safely and efficiently, these systems help streamline workflows while protecting product quality and reducing contamination risk.
Thermo Fisher Scientific provides a comprehensive selection of standard and customized fluid transfer products. Our offerings include the dependable and leakproof Thermo Scientific Nalgene fluid transfer tubing as part of our Thermo Scientific standard line of fluid transfer assemblies with the BioTitan Retention Device that connect to a variety of bioprocess bags and bottles. Additionally, many of our products can be easily configured using our Single-Use Assembly Configurator tool.
Our Single-Use Assembly Configurator Tool makes it easy to create custom assemblies tailored to your process or select in-stock, ready-to-ship options to reduce lead times. Every design is quickly reviewed, with rapid quotations and detailed 3D renderings to ensure it meets your exact requirements before production begins.
We offer customized fluid transfer solutions designed specifically for the unique needs of your process. Leverage our solutions to help reduce the risk of sterility failure and save on sterilization costs. Supports “lean” systems by decreasing inventory, simplifying supply chains, and reducing labor costs.
Experience fast lead times when leveraging standard products for your fluid transfer needs. In addition, we offer a large portfolio of stocked and ready to ship fluid transfer products to support your bioprocessing applications.
Fluid transfer solutions for bioprocessing include single-use tubing, connectors, bags, bottles, containers, and assemblies that can integrate with bioreactors, mixers, and cell factories. Our solutions include the BioTitan Retention Device which is designed to form a 360-degree seal to prevent leaks.
There are many benefits of fluid transfer solutions including the reduction of leaks and spills reducing contamination risk. They provide consistency of handling and are particularly useful for the movement of fluids in and out of large or difficult to access vessels like bioreactors.
Fluid transfer solutions support closed system bioprocessing by offering sterile, leakproof connections that help minimize the risk of contamination. They enable easy integration with bioreactors, mixers, and other equipment. The BioTitan Retention Device helps improve the integrity of closed systems by forming a 360-degree seal around tubes and fittings.
Standard assemblies are stocked and can be ordered directly with relatively short lead times. We offer several types of custom assemblies that have different lead times. Some assemblies can be configured from in stock components using the Single-Use Assembly Configurator. More customized solutions can be made from components within our Global Standard Component Library (GSCL).
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.