Search
(A) Amplification results are shown using TaqPath 1-Step Multiplex Master Mix (No ROX) over 6 logs for a quadplex reaction using human cDNA and assays for CD44 (red), CYC1 (green), TMSB10 (orange) and G6PD (blue) (all with R2 = 1.0). (B) Amplification results are shown using TaqPath 1-Step Multiplex Master Mix (No ROX) over 6 logs for a quadplex reaction using human RNA and assays for CD44 (red), CYC1 (green), TMSB10 (orange) and G6PD (blue) (all with R2 = 1.0).