Thermo Fisher Scientific  ›  Electron Microscopy   Electron Microscopes     3D Visualization, Analysis and EM Software   Customer Portal
PerGeos and Avizo Software at InterPore 2023
The 15th Annual International Conference on Porous Media will range from pore-scale modelling, pore-scale imaging, to experimental and numerical methods on larger scales.
May 22-26, 2023 | Edinburgh, Scotland | Booth # 10
PerGeos Software and Avizo Software are all-in-one digital labs that allow for the visualization, processing, and quantification of porous materials. Our analytical software enables the detection and classification of various types of porosity, even on images with complex artifacts. Many porosity properties and statistics can be calculated, such as volume fraction, largest ball fitting through a given pore, pore size distribution, pore throat size distribution, pore orientation, shape factor, and other porosity properties. Porosity can be turned into a model (Pore Network Model), allowing for rapid understanding and exploration of the pore space that features spheres or ellipsoids-and-sticks type visualization with property mapping. PerGeos Software and Avizo Software also allow the direct calculation of the absolute permeability of the material from the segmented pore space.
Don’t miss our short course on 3D imaging of porous materials (e.g., rocks, batteries).
>>Discover >