Amira-Avizo 3D Software Download Page
As part of our ongoing effort to improve your experience, this page is no longer being updated and all future software release notes and update information will now be available on our new, user-friendly portal: Software Resources and Updates for Electron Microscopy Instruments
IMPORTANT: Make sure to have administrator rights in order to install Amira-Avizo.
The following applications are included in the Amira-Avizo installers:
– Windows: Amira3D, Avizo3D, Avizo for Industrial Inspection, ToGo Viewer Software for Amira and Avizo Software, SMSConverter
These applications run on: Microsoft Windows® 10 and Microsoft Windows® 11 (64-bit). Testing on Windows® versions older than Windows® 10 has been discontinued.
– Linux: Amira3D, Avizo3D, ToGo Viewer Software for Amira and Avizo Software, SMSConverter
These applications run on: Linux® x86_64 (64-bit). Supported 64-bit architecture is Intel64/AMD64 architecture. Supported Linux distribution is Ubuntu 20.04 (64-bit PC desktop image).
You will find complete details about Amira-Avizo Software system requirements in Amira-Avizo Software system requirements.
About Amira-Avizo3D 2025.1.1 License Management
You will find complete details about Amira-Avizo licensing and activation in Amira-Avizo Software License Manager.
Flexnet Server
If you are using Floating Licenses, managed by a FlexNet License Server, make sure to have installed version 11.19 or higher of the FlexNet tools on your system.
For more details and download links, please refer to: FlexNet Floating License Server Documentation and Utilities Download Links
You will find complete install/uninstall instructions in Amira-Avizo software install uninstall instructions.
ToGo Viewer Software for Amira and Avizo Software Publisher extension is included in Avizo Software 3D Pro, Amira Software 3D Pro, Avizo Software 3D for Industrial Inspection and Amira Software 3D for Cell Biology. This extension allows you to publish the results of your analysis using a special Avizo Software Data Format with all the project data in a single file. This file can then be opened by the free ToGo Viewer Software, allowing you to share the dataset in an interactive way with colleagues and customers who do not have a full version of Amira-Avizo Software. For more information, read ToGo Viewer Software for Amira and Avizo Software description.
Previous versions
Amira-Avizo 3D 2024.2 for Microsoft Windows
Download Amira-Avizo3D 2024.2 for Microsoft Windows 64-bit (3.8 GB)
Amira-Avizo 3D 2024.2 for Ubuntu (20.04) Linux
Download Amira-Avizo3D 2024.2 for Ubuntu Linux 64-bit (5.39 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira-Avizo 3D 2024.1 for Microsoft Windows
Download Amira-Avizo3D 2024.1 for Microsoft Windows 64-bit (2.97 GB)
Amira-Avizo 3D 2024.1 for Ubuntu (20.04) Linux
Download Amira-Avizo3D 2024.1 for Ubuntu Linux 64-bit (2.61 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira-Avizo 3D 2023.2 for Microsoft Windows
Download Amira-Avizo3D 2023.2 for Microsoft Windows 64-bit (3 GB)
Amira-Avizo 3D 2023.2 for Ubuntu (20.04) Linux
Download Amira-Avizo3D 2023.2 for Ubuntu Linux 64-bit (2.7 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira-Avizo 3D 2023.1.1 for Microsoft Windows
Download Amira-Avizo3D 2023.1.1 for Microsoft Windows 64-bit (3.21 GB)
Amira-Avizo 3D 2023.1.1 for Ubuntu (20.04) Linux
Download Amira-Avizo3D 2023.1.1 for Ubuntu Linux 64-bit (2.47 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira-Avizo 3D 2022.2 for Microsoft Windows
Download Amira-Avizo3D 2022.2 for Microsoft Windows 64-bit (3.4 GB)
Amira-Avizo 3D 2022.2 for Ubuntu (20.04) Linux
Download Amira-Avizo3D 2022.2 for Ubuntu Linux 64-bit (2.6 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira-Avizo 3D 2022.1 for Microsoft Windows
Download Amira-Avizo3D 2022.1 for Microsoft Windows 64-bit (3.3 GB)
Amira-Avizo 3D 2022.1 for CentOS (RHEL) Linux
Download Amira-Avizo3D 2022.1 for CentOS Linux 64-bit (2.4 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira 2020.3 for Microsoft Windows
Download Amira 2020.3 for Microsoft Windows 64-bit (2.6 GB)
Amira 2020.3 for Red Hat Enterprise Linux
Download Amira 2020.3 for CentOS Linux 64-bit (2.1 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Amira 2020.3 for OS X
Download Amira 2020.3 for OS X 64-bit (2.0 GB)
You need to make sure that your security settings allow installation of Amira, which does not come through the Apple Store. To allow installation of Amira, just right-click on the installer and select Open.
If the package installation fails, please open a terminal window, navigate to the directory where you downloaded the installer, then run the command:
sudo installer -allowUntrusted -pkg "Amira-202031-MacOSX-clang70.pkg" -target /
Avizo 2020.3 for Microsoft Windows
Download Avizo 2020.3 for Microsoft Windows 64-bit (2.3 GB)
Avizo 2020.3 for Red Hat Enterprise Linux
Download Avizo 2020.3 for CentOS Linux 64-bit (2.2 GB)
If the download doesn’t start when clicking on the above link, please right-click on it and select “Save link as…” to save the installer in the directory of your choice.
Avizo 2020.3 for OS X
Download Avizo 2020.3 for OS X 64-bit (2.2 GB)
IMPORTANT: You need to make sure that your security settings allow installation of Avizo, which does not come through the Apple Store. To allow installation of Avizo, just right-click on the installer and select Open.
If the package installation fails, please open a terminal window, navigate to the directory where you downloaded the installer, then run the command:
sudo installer -allowUntrusted -pkg "Avizo-202031-MacOSX-clang70.pkg" -target /