Thermo Fisher Scientific  ›  Electron Microscopy   Electron Microscopes     3D Visualization, Analysis and EM Software   Customer Portal
New XTra: Dual Energy CT (DECT)
Xtras are add-ons and how-to videos that will help you improve your daily use of Amira and Avizo Software.
Add on: Advanced voxel-by-voxel density and atomic number calculation for enhanced whole-core CT analysis
The DECT-4 add-on allows for the calculation of density and effective atomic number on a voxel-by-voxel basis using dual-energy X-ray CT data. This technology enhances whole-core CT analysis by providing detailed density information that can be compared with density logs and used for further rock formation characterization. The add-on includes several modules and a calibration recipe to support comprehensive analysis workflows.
>> Discover add on
>> Discover the Xtra Library