Thermo Fisher Scientific  ›  Electron Microscopy   Electron Microscopes     3D Visualization, Analysis and EM Software   Customer Portal
New XTra: Trabecular Segmentation Workflow for Murine Tibial Bone
Xtras are add-ons and how-to videos that will help you improve your daily use of Amira, Avizo and PerGeos Software. Each month, we will highlight one of the newest and greatest Xtra for your best productivity!
Add on: Trabecular Segmentation Workflow for Murine Tibial Bone
This Xtra offers a workflow using watershed and thresholding tools to segment trabecular bones from the epiphysis region in murine tibial bone. It includes a method for generating markers and a recipe for trabeculae segmentation.
>> Discover add on
>> Discover the Xtra Library