Amira-Avizo Software Install/Uninstall Instructions Page
To access Amira-Avizo install/uninstall instructions, please go to our Software Resources and Updates for Electron Microscopy Instruments Portal
Installation
Standard Installation
Follow the instructions below for your operating system.
Microsoft Windows
When the download is finished, double-click on the Amira-Avizo Software installer and follow the instructions. Note that administrator rights are required.
In the Select Components section, specify the desired installation type:
  • Typical installation installs the standard component of the software.
  • Developer installation adds developer files and binaries files for debugging (Amira 3D Pro Edition is needed).
  • Custom installation lets you choose the component to install.
After selecting the installation type, follow the Amira-Avizo Software installation instructions.
Linux
When the download is finished, go to the directory where the Amira-Avizo Software installer is and open a terminal window.
  1. Add execution rights to the file ./Amira-XXX-Linux64-gccYY.bin by entering chmod a+x ./Amira-XXX-Linux64-gccYY.bin in the command line.
  2. Enter: sudo ./Amira-XXX-Linux64-gccYY.bin (XXX denotes the product version of Amira and YY the compiler version) in the command line.
  3. Click Next and press Enter.
  4. In the Select Destination Folder, choose the Amira-Avizo Software installation destination.
  5. In Select Components, choose components:
    • Main / Runtime files to install Amira-Avizo Software
    • Developer files to install developer files and binaries files for the developer to debug
      Note: You must have an Amira-Avizo Software 3D Pro Edition for this option.
  6. Click Next and press Enter to finalize the installation.
  7. You can now launch the software with this command:
    /usr/local/AmiraAvizo3D/XXX/bin/Amira3D
    or /usr/local/AmiraAvizo3D/XXX/bin/Avizo3D
    Adapt the path if you have not used the default path.
  8. At this point, you may have an error regarding a missing license as the product has not been activated.
    Refer to the procedure provided in your delivery email or our online documentation for the steps to follow depending on your license.
Silent Installation
Amira-Avizo Software allows silent installation via the command line on both Windows and Linux. Silent installation installs the software with the default parameters.
Note that there is no option to automate the floating licensing setup on client machines, but you can easily create the client license file and the environmental variable ASGLMD_LICENSE_FILE that points to this file.
For example, you can create the license file in the Avizo folder “share\license” under the name license.dat with these lines:
SERVER <server_hostname> ANY <port_number>
USE_SERVER
Where:
  • <server_hostname> is the hostname or IP address of your server machine,
  • <port_number> is the TCPIP port used by the server Imadmin/Imgrd for communicating with client machines (be careful to configure the firewall on each client machine for opening this port.)
Windows
To install Amira-Avizo Software as a silent installation:
  1. Open a Windows command prompt as administrator (This procedure does not work from an Unix shell).
  2. Navigate to the Amira installer folder.
  3. In the command prompt, enter:
    Amira-XXX-Windows64-VCYY.exe /VERYSILENT /SUPRESSMSGBOXES /NORESTART /NOCANCEL /KEEPEXISTINGPATHPOLICY  /DIR=installPath
    Note: XXX denotes the product version of Amira-Avizo Software and YY the compiler version.
Note that the option /KEEPEXISTINGPATHPOLICY permanently disables the Windows maximum path limit (260 characters) and requires administrator privileges. You can remove this command if you do not want to disable this limit.
The installation begins after the line is entered.
Silent installation does not install the Thermo Fisher License Manager. You can find the TLM installer in the folder share\license\TLM located in the Amira-Avizo Software installation directory for manual installation.
Linux
With the silent argument, the installer is in silent mode and does not ask you for any confirmation or question. All needed information is expected in the command line.
Open a terminal and run the Amira-Avizo Software installer with the following command line:
./Amira-XXX-Linux64-gccYY.bin — –silent –eula-ok –dir install_dir
Command line explanations follow.
  • Use the double dash  with no keyword preceding the entire set of arguments.
  • If you are in the silent mode, the EULA must be accepted by using mandatory command-line argument –eula-ok.
  • Modify the installation directory with the command-line argument –dir install_dir, so that the product is unpacked in the install_dir / product / version directory. You can set absolute path or path relative to installer script location.
  • By default, the silent installer only unpacks the runtime of Amira-Avizo Software. For the developer installation, add command-line argument –dev-install.
Uninstallation
To uninstall Amira-Avizo Software from your machine:
  • On Microsoft Windows, remove Amira-Avizo Software with the standard Add or remove programs
  • On Linux open a terminal and run this command line, replacing install_dir with the location of the Amira-Avizo Software install:
rm -rf install_dir
CAUTION: This command irretrievably removes all contents of the chosen folder.
Remove custom Python environments
With Amira-Avizo Software, you can install custom Python environments via the Python menu. If you wish to delete these environments, on Microsoft Windows they are stored in the folder C:\Users\UserName.edm\envs and on Linux in the folder $HOME/.edm.
Remove custom elements
If you have installed custom elements, such as add-ons from the Xtra Library, you may also wish to remove these from your system. The default installation directory for Xtras on Windows is %APPDATA%\Thermo Fisher Scientific\Xtras\Amira-Avizo\<Amira-Avizo-Version>. On Linux, the directory is /home/<username>/.config/Thermo Fisher Scientific/Xtras/Amira-Avizo/<Amira-Avizo-Version>.
ToGo Viewer Software for Amira and Avizo Software Publisher extension is included in Avizo Software 3D Pro, Amira Software 3D Pro, Avizo Software 3D for Industrial Inspection and Amira Software 3D for Cell Biology. This extension allows you to publish the results of your analysis using a special Avizo Software Data Format with all the project data in a single file. This file can then be opened by the free ToGo Viewer Software, allowing you to share the dataset in an interactive way with colleagues and customers who do not have a full version of Amira-Avizo Software. For more information, read ToGo Viewer Software for Amira and Avizo Software description.