Module: AddDataToPointCloud ()
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.AddDataToPointCloud writes data defined on mesh-nodes into a point-cloud with equal number of points, e.g. the result of applying Extract Point Cloud to the mesh.
Data [required]
The point-cloud.Data2 [optional]
The data defined on the mesh-nodes, i.e. data of type HxSurfaceScalarField or HxSurfaceVectorField.
The number of values has to be equal to the numbers of points in the point-cloud.