Module: CopyMeasureAnnotationSettings ()
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.This Tcl script-object can be used to copy visualization settings from one measure to all others in a Measurement module. It is also possible to save/load settings to/from a text file.
Mesurement instance [required]
The selected Measurement module.
Reference measure index
Use the slider to select the measure whose settings shall be copied to all the others.
Measure label
The label of the selected measure.
filename
Specify a file name here, for saving or retrieving settings.
Action