Module: ExportMeasures ()
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
It might be desired to export the measures and annotations created with the Measurement tool of Amira/Avizo for further analysis.
This Tcl script-object creates a spreadsheet with the data of the measurements and annotations.
Connect this Tcl script-object to a Measurement module.
On "Apply", a spreadsheet with the data of the objects is created.
The spreadsheet can then be exported, e.g. to CSV or XML, or its contents can be selected and copied to the clipboard and then pasted e.g. into Excel.
Mesurement instance [required]
The Measurement module, whose data you want to export.