Module: splitIntoBlocks-recipeForEachBlock ()
This is an Xtra TCL script that splits a binary pore space into blocks and output one porosity and permeability value for every block.Choose the appropriate binary pore space in the data port, and indicate how many blocks you want in X, Y and Z. Check if you need both porosity and permeability to be computed for each block, and press Apply.
After the computation is done, you get 3 outputs :
- a spreadsheet with the block descriptions and the computed values
- the porosity by block data
- the permeability by block data
Data [required]
Input Binary Pore Space.
dividerx
Number of sub-divisions in direction Xdividery
Number of sub-divisions in direction Ydividerz
Number of sub-divisions in direction ZOptions
Select recipes to be executedRecipe1
First recipe (default = porosity)Recipe2
First recipe (default = permeability)