Module: Label to Area ()
Description:
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
This module converts a binary image into a grayscale image where the value of all non-empty voxels equals the area of the binary label input.
The resulting grayscale image can then be used to compute the area fraction of any sub-area against the area of the binary label.
Connections:
Data [required]
Input image.
Ports:
Console
Opens the Python Script Object console of the module as the active console window.