Module: Scale Axes ()
Description:
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
The coordinate-axes displayed using the "Axes" module cannot easily be scaled. Using this Xtra, you can easily change the lengths of the axes and the location of the displayed origin.
You can change the length of each displayed axis separately, using the sliders in the module's properties. It is also possible to move the origin of the displayed axes. If the allowed value-range is not enough, you can adjust it after pressing the "..." buttons.
Note: "Axes" is automatically changing the thickness of the displayed axes at certain length values. You can change that using the "Thickness" slider in "Axes"'s properties.
Connections:
Axes module [required]
The "Axes" module to change the axes' properties of.
X length
The displayed length of the X-axis.
Y length
The displayed length of the Y-axis.
Z length
The displayed length of the Z-axis.
X origin
The X-position of the origin of the displayed axes.
Y origin
The Y-position of the origin of the displayed axes.
Z origin
The Z-position of the origin of the displayed axes.