Module: Crop Point Cloud ()
Description:
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
This module crops a point cloud object (HxCluster) according to a ROI-Box. Data and Label columns defined on the source point cloud will be copied accordingly into the cropped point cloud. Connect port ROI Box with the ROI-Box and click Apply.
Connections:
Data [required]
This port takes a Point Cloud object HxCluster.
ROI Box [optional]
Connect this port with the ROI-Box defining the region-of-interest.