Convex Hull 3D
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
This module generates the 3D convex hull of a volume from a binary label image.
Connections
Input
Description
Data: HxUniformLabelField3
Binary label image representing the volume from which to compute the 3D convex hull of
Outputs
The module generates a binary label image representing the computed 3D convex hull.