Module: ChannelSelector ()
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.This module simply duplicates a channel from a MultiChannelField object. This is only useful for creating recipes that can operate on such objects.
Data [required]
MultiChannelField data object.
Select Channel
Index of the channel to be selected.