Module: Thinner2d ()
Description:
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
This module applies the Zhang-Suen thinning algorithm slice by slice to a binary image until only a string of connected voxels remains.
Connections:
Data [required]
The binary image to be thinned.