Thinner2d
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
This module applies the Zhang-Suen thinning algorithm slice by slice to a binary image until only a string of connected voxels remains.
Connections
|
Input |
Description |
|---|---|
|
Data: [required] |
The binary image to be thinned. |