Enlarge
This is an experimental module from the Xtra Library: www.thermofisher.com/software-em-3d-vis/xtra-library/.This Xtra module enlarges the data in the specified directions and by a specified number of pixels. The value assigned to the added pixels is 0 by default and can be set to another value. A Replicate mode also allows to assign, in a given direction, the values of the border slice of the input data to the new layer slices.
Connections
|
Input |
Description |
|---|---|
|
Data: [required] |
The input data should be a uniform scalar or label field. |
Parameters (Ports)
|
Parameter |
Description |
|---|---|
|
Adjust min by [px]: |
Specify the width in pixels of the layers added in each direction. |
|
Adjust max by [px]: |
Specify the width in pixels of the layers added in each direction. |
|
Pixel value: |
Value assigned to the newly created pixels. If Replicate is selected, then the value can no longer be set, and the values of the border slice of the input data are assigned to the new layer slices. |