Flip and Swap
This is an experimental module from the Xtra Library:
www.thermofisher.com/software-em-3d-vis/xtra-library/.
This Xtra module flips or swaps uniform scalar or label fields in specified directions. Note that applying several time the module will only apply the latest transformation to the input data and overwrite the result. To cumulate transformations, a sequence of several modules needs to be created.
Connections
|
Input |
Description |
|---|---|
|
Data: [required] |
The input data should be a uniform scalar or label field. |
Parameters (Ports)
|
Parameter |
Description |
|---|---|
|
Flip: |
Reverts the order of slices in the corresponding dimension. |
|
Swap: |
Interchanges the corresponding dimensions. |