Module: Project Spatial Graph ()
Description:
This is an experimental module from the Xtra Library: https://xtras.amira-avizo.com.
This module projects a Spatial Graph on a user-defined plane.
The plane can be defined manually or by connecting a plane (e.g. a Slice, Clipping Plane) or by connecting a spreadsheet containing barycenter and eigenvectors information (e.g. Label Analysis).
Connections:
Data [required]
The Spatial Graph to be projected.
Plane [optional]
A planar object on which to project the Spatial Graph (e.g. Clipping Plane).
Analysis [optional]
A spreadsheet containing barycenter and eigenvectors information (e.g. Label Analysis) from which a plane can be defined.
Ports:
Plane Definition
Pick an option for plane definition. Plane and Analysis can be chosen only if a plane or spreadsheet are connected from the eponym ports.
Row Index
This port is displayed if Analysis is selected and if the spreadsheet has more than one row. In that case, you can select the row from which the plane will be defined.
Origin
Origin of the plane.
Note: This port can't be edited if Plane and Analysis are chosen. To edit a plane defined from e.g. a Slice, go back to Manual definition and edit.
Plane Vec 1
First vector defining the plane.
Note: This port can't be edited if Plane and Analysis are chosen. To edit a plane defined from e.g. a Slice, go back to Manual definition and edit.
Plane Vec 2
Second vector defining the plane.
Note: This port can't be edited if Plane and Analysis are chosen. To edit a plane defined from e.g. a Slice, go back to Manual definition and edit.
Create Plane
Allows the creation of an Empty Plane from the plane definition information stored in the module.