Which product to choose depends on your ligand, target, sample and application. Both 1 micron (MyOne), 2.8 micron (M-270/M-280) and 4.5 micron (M-450/M-500) Dynabeads magnetic beads can be used. If the sample is very viscous, 4.5 micron beads are recommended. If the target is very fragile, smaller beads are recommended, as they will move more slowly towards the magnet, reducing the risk of squeezing the target. Below is a short product guide showing which beads can be used to immobilize different ligands:
For antibody ligands, use anti-mouse, anti-rat or anti-rabbit coated beads or Dynabeads magnetic beads Protein A/G (2.8 micron beads), Dynabeads magnetic beads M-280 Tosylactivated, Dynabeads magnetic beads M-450 Epoxy or Dynabeads magnetic beads M-500 Subcellular.
For biotinylated ligands, use Dynabeads magnetic beads M-280 Streptavidin. The hydrophilic, negatively charged Dynabeads magnetic beads M-270 Streptavidin or Dynabeads magnetic beads MyOne Streptavidin C1 may also be used.
For protein/peptide ligands (other than antibodies), use Dynabeads magnetic beads M-270 Epoxy, Dynabeads magnetic beads M-270 Amine, Dynabeads magnetic beads M-450 Epoxy.
For histidine affinity-tagged ligands, use Dynabeads magnetic beads His-Tag Isolation and Pulldown beads (1 micron size).
Find more information about Dynabeads magnetic beads at
http://www.thermofisher.com/Dynabeads magnetic beads.
Find additional tips, troubleshooting help, and resources within our
Protein Assays and Analysis Support Center.