KnockOut™ Serum Replacement, 500 mL - FAQs
View additional product information for KnockOut™ Serum Replacement - FAQs (10828028, 10828010)
2 product FAQs found
Can KnockOut SR be used to culture mouse ESCs in feeder-free conditions?
Yes, mouse embryonic stem cells can be cultured using KnockOut SR under feeder-free conditions. Typically, cells are plated at a higher seeding density, in the presence of LIF, on a 0.1% gelatin layer in place of feeder cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can KnockOut SR be used to culture mouse ESCs on feeder cells?
Yes, KnockOut SR was designed to culture mouse ES cells in undifferentiated conditions.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.