Search
View additional product information for 2-Mercaptoethanol - FAQs (21985023)
1 product FAQs found
The 2-Mercaptoethanol must be added to the medium immediately before use because activity rapidly declines in diluted form. You can supplement enough medium needed for that day.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.