Search
View additional product information for Sulfo-SBED Biotin Label Transfer Reagent, No-Weigh™ Format - FAQs (33034)
10 product FAQs found
Sulfo-SBED allows one to transfer a biotin tag from an amine-containing protein to a second protein (reacted with the phenyl azide). The amine-containing protein can be removed by reduction of the disulfide in the Sulfo-SBED spacer arm. This leaves just the second biotinylated protein for detection.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
During the initial coupling procedure, the solutions should be protected from light to avoid decomposition of the phenyl azide functional group.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
The NHS-ester end of the crosslinker is subject to hydrolysis, therefore the amine-containing protein should be conjugated prior to the photoactivatable (aryl azide) reaction.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
UV light at wavelengths greater than 300 nm are appropriate for conjugation.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Yes, Sulfo-SBED is soluble in aqueous as well as organic solutions. It is most soluble in organics such as DMF and DMSO.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
No, the crosslinker is moisture sensitive. The sulfo-NHS ester is easily hydrolyzed.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
In the case of sulfo-SBED biotin, there is a disulfide in the spacer arm of this tri-functional crosslinker that is cleaved with reducing agents such as DTT, beta-ME or TCEP. As the disulfide lies between the contact point on the bait protein and the complex of the photoreactive group and biotin, reduction separates the bait protein from the prey protein; which is now biotin labeled.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
The NHS-ester of sulfo-SBED biotin will react with primary amines on the bait protein. The photoreactive aryl azide primarily targets nucleophiles, such as -NH2, -SH and -OH, but in their absence, it will react with any near neighbor molecule.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Currently, we offer sulfo-SBED Biotin Label Transfer Reagent (Cat. Nos. 33033 and 33034) as well as the Sulfo-SBED Biotin Label Transfer Kit for Western Blot Application (Cat. No. 33073).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.