Silica gel orange, for drying purposes, non toxic grade, 2-5 mm - FAQs
View additional product information for Silica gel orange, for drying purposes, non toxic grade, 2-5 mm - FAQs (392030050, 392030010, 392032500)
2 product FAQs found
How can I determine if there is moisture uptake by the Silicagel orange, for drying purposes, non toxic grade?
The degree of moisture uptake will be indicated by color change from orange to colorless at approximately 6 wt.% water content. However, it continues to adsorb water from this point until reaching its maximum capacity.
What is the recommended protocol for regeneration of Silicagel orange, for drying purposes, non toxic grade?
Silicagel orange, for drying purposes, non toxic grade full adsorption capacity can be regained by regeneration at 90 degrees Celsius until its original orange color is obtained.