Search
View additional product information for Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da - FAQs (436200050, 436200010)
1 product FAQs found
Our Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da is acid-terminated.