Cyclosporin A, 98% - FAQs
Cyclosporin A, 98% - FAQs
View additional product information for Cyclosporin A, 98% - FAQs (457970050, 457970010)
1 product FAQs found
Which solvents can I use to dissolve cyclosporin A, 98%?
Cyclosporin A is soluble in ethanol, chloroform, methylene chloride/dichloromethane, and DMSO.