LB Broth, Lennox - FAQs
View additional product information for LB Broth, Lennox - FAQs (612725000)
2 product FAQs found
What is the composition of LB Broth, Lennox (Cat. No. 612725000)?
LB Broth, Lennox (Cat. No. 612725000) is composed of 10.0 g/L casein digest peptone, 5.0 g/L sodium chloride, and 5.0 g/L yeast extract.
How should I prepare LB Broth, Lennox (Cat. No. 612725000)?
We recommend the following:
1. Mix 20 grams of the LB broth powder in 1 liter of purified water.
2. Agitate to completely dissolve.
3. Autoclave for 15 minutes at 121°C.