What is your recommended method for thawing L-Glutamine before adding to cell culture media?
We recommend thawing at 2-8 degrees C and then warming at 37 degrees C until the material goes into solution. Mix it as it warms up to spend minimal time at elevated temperatures before you aliquot it.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.