Search
Buscar
View additional product information for TrueGuide™ gRNA - FAQs (A35512)
1 product FAQs found
The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.