Neurobasal™ Plus Medium - FAQs
Neurobasal™ Plus Medium - FAQs
View additional product information for Neurobasal™ Plus Medium - FAQs (A3582901)
14 product FAQs found
Can Neurobasal Medium alone or Neurobasal Medium supplemented with B-27 supplement be frozen to extend product shelf life?
No, it is not recommended to freeze media due to the potential of precipitates forming upon thaw. Inorganic salts and amino acids in the formulation may come out of solution when exposed to temperature fluctuations. These precipitates will not go back into solution easily once formed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which media system should I use for primary culture of P0- or P1-derived mixed hippocampal cells?
We recommend using Neurobasal Plus Medium with B-27 Plus Supplement for long-term culture of mixed hippocampal cells. For short-term culture of a pure population of hippocampal neurons, we would recommend using Neurobasal-A Medium with B-27 Supplement.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the osmolality of Neurobasal Plus Medium?
The osmolality is in the range of 215-235 mOsm/kg. The exact osmolality for your lot of the medium can be found in the Certificate of Analysis for that lot number.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?
We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?
We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I thaw B-27 Supplements?
We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I use B-27 Plus Neuronal Culture System to culture non-neuronal cells?
The B-27 Plus Neuronal Culture System was designed for culturing neuronal cells. We have not tested it in other cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I am planning to use the B-27 Plus Neuronal Culture System. How can I control the number of glial cells in my neuronal culture?
Add CultureOne Supplement at day 0 with the B-27 Plus Neuronal Culture System to fully suppress both astrocytes and oligodendrocytes with no detrimental effect on neurons. Delaying the addition of CultureOne Supplement to later time points results in increasing levels of astrocytes. Refer to the CultureOne Supplement User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0016204_CultureOneSupplement_UG.pdf) for more information.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do I have to use B-27 Plus Supplement with Neurobasal Plus Medium?
B-27 Plus Supplement and Neurobasal Plus Medium have been designed to work synergistically together for maximal performance. They can be used with other basal media or supplements if necessary.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How long can I maintain my neurons in culture with the B-27 Plus Neuronal Culture System?
We have maintained primary rat cortical neurons up to 8 weeks and rat hippocampal neurons up to 4 weeks. Mouse cortical neurons were maintained for 4 weeks. Human iPSC-derived neurons that have been differentiated from neural stem cells (NSCs) for 9 days have been maintained for 5 weeks. All cells tested with the B-27 Plus System were significantly healthier than cells maintained in culture using the classic products across all time points.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I use the B-27 Plus Neuronal Culture System for expansion of proliferating cells, such as neural stem cells (NSCs)?
No, we do not recommend the B-27 Plus System for expanding proliferating cells. We recommend using StemPro NSC SFM, Neural Induction Medium Supplement or B-27 Supplement minus Vitamin A for growing proliferative cells, such as NSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I re-freeze B-27 Supplements into smaller aliquots for future use?
Yes, we recommend that you aliquot when you first thaw it and thereafter subject the aliquot to no more than 2 additional freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How did you upgrade the manufacturing/QC process of the B-27 Plus Supplement and Neurobasal Plus Medium?
We streamlined the manufacturing process, implemented more stringent acceptance criteria and enhanced QC assays for qualification of raw materials and release of finished product to enable better lot-to-lot consistency.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Neurobasal Plus Medium and Neurobasal Medium?
Key amino acids and buffering components have been optimized in Neurobasal Plus Medium. No small molecules or growth factors have been added.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.