Salmon Sperm DNA, sheared (10 mg/mL) - FAQs
View additional product information for Salmon Sperm DNA, sheared (10 mg/mL) - FAQs (AM9680)
3 product FAQs found
How many freeze thaw cycles do you recommend for Invitrogen, Salmon Sperm DNA, sheared (Cat. No. AM9680)?
We do not have specific guidance on the number of freeze thaws for Salmon Sperm DNA, but it can be frozen and thawed multiple times.
Do you have an alternative for Salmon Sperm DNA, sheared (10 mg/mL) (Cat. No. AM9680) to use as a spike-in for standard curve preparation for ssDNA?
You can use Yeast tRNA (10 mg/mL) (Cat. No. AM7119). When used at a final concentration of 10–20 µg/mL, yeast tRNA is an effective coprecipitant to aid in recovery of small amounts of nucleic acids.For more information, see the following link.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
What is the molecular size of Salmon Sperm DNA (Cat. No. AM9680)?
The molecular size of the Salmon Sperm DNA (Cat. No. AM9680) is around 200-500 bp.