Search
View additional product information for Human Complement C3a ELISA Kit - FAQs (BMS2089TEN, BMS2089TWO, BMS2089, BMS2089FIVE)
18 product FAQs found
You can find expected "normal levels" listed in the Platinum ELISA Kit manual under "Expected Values". Please be aware that these normal values were obtained in our laboratories with a limited number of samples. We strongly suggest that each laboratory perform studies in order to establish normal range for their sample populations.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
In case of small reagent volumes, please spin down vial before use to make sure to all contents are collected in the bottom of the tube.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
We recommend following the instructions for sample dilutions as provided in the manual. We cannot guarantee optimal performance of the assay if internal assay dilutions as instructed were omitted.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
Incubation times should be followed exactly as stated in the product manual to ensure optimal Platinum ELISA test performance.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
An initial washing step is not necessary but recommended as accuracy may be increased.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
We recommend that you use the manual included with the Platinum ELISA kit.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
The reagents within the Platinum ELISA kits are not interchangeable between different lots of the same kit or between kits against different analytes.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
No, but we strongly recommend that you make at least double determinations of each sample and standard point.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
Most ELISAs are suitable for serum, plasma, cell culture supernatants and other body fluids. Detailed information is given in the product manual for each individual kit.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
All reagents necessary to perform 1 or 10 plate ELISA assays, including optimized capture-antibody pre-coated plates.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
The supernatants can be harvested and stored -20 degrees C. Avoid freeze/thaw cycles. We recommend that you briefly centrifuge samples after collection to pellet any dead/floating cells remaining in the supernatant. Transfer to new tube for storage.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
Each Ready-Set-Go! ELISA Set comes with its own Certificate of Analysis (CofA). The directions on the CofA should be strictly followed to ensure performance of the reagents. The CofA gives specific instructions on how to dilute each component of your lot specific set. The instructions on the CofA should not be deviated from in order to ensure accurate results.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
The calculation factor is always stated in the manual.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
For some Instant ELISA kits, the reconstitution volume of the standard and blank wells may differ (these components are lot specific). Please use the manual provided in the kit for specific volumes to use.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
In general, you can use undiluted samples with the Instant ELISA. Please refer to the product manual to determine if dilution of the sample is required for your kit of interest. The volume of sample required for the individual kits will also be listed in the product manual.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
No, the secondary antibody is included as a component of the lyophilized reagents in the wells.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
No, the recombinant standard is included as a component of the lyophilized reagents in the wells.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
Performance of Platinum ELISA cannot be guaranteed if reagents are not stored as indicated.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.