iCAP™ RQ ICP-MS - FAQs
View additional product information for iCAP™ RQ ICP-MS - FAQs (BRE731416)
26 product FAQs found
What systems are available for speciation analysis?
Any of the Thermo Scientific Dionex IC systems can be used in conjunction with the iCAP RQ ICP-MS system. Visit our site to see all system configurations (https://www.thermofisher.com/us/en/home/industrial/chromatography/ion-chromatography-ic/ic-icp-ms-speciation-analyzer.html)
What example analytes require speciation analysis?
The following are some of the most common analytes screened by speciation analysis:
• Arsenic
• Chromium
• Iodide
• Mercury
Visit our Speciation Analysis by IC-ICP-MS page to view our application note videos where our scientists walk through speciation analysis of example analytes:
• Speciation Analysis of Arsenic in Organic Brown Rice Syrup using the Thermo Scientific iCAP-Q ICP-MS
• Chromium Speciation Analysis in Drinking Water by IC-ICP-MS
• Analysis of Arsenic Species in Apple Juice by IC-ICP-MS
How is speciation analysis performed?
IC-ICP-MS provides a complete inorganic elemental analysis solution for speciation analysis.
• Metal-free ion chromatography (IC) separates the individual ionic species without contributing trace metal contamination
• Ion chromatography-inductively coupled plasma mass spectrometry (IC-ICP-MS) performs trace elemental detection and quantification
Metal-free IC (with high resolution ion exchange columns and simple online connectivity) together with high sensitivity ICP-MS and integrated software are a powerful combination for fast and efficient metal speciation. Visit our Speciation Analysis by IC-ICP-MS page to see our scientists discuss real-life examples and benefits of combining IC and ICP-MS workflows for speciation analysis.
Why is speciation analysis important?
Speciation analysis has become important for food, environmental, and pharmaceutical industries, where simply measuring the total amount of an element is insufficient. Identifying the different species and concentrations of those species provides a more informed understanding of the environmental or health related impact associated with a particular sample. The clinical or environmental approach would be very different depending on the concentration of the toxic species. For example, an exposure to toxic inorganic arsenic (arsenite and arsenate) is a much bigger concern than an exposure to the mildly toxic organic arsenic species. In contrast, organic mercury species are more toxic than inorganic mercury species, so an exposure to methylmercury or ethylmercury poses a more severe health risk.
What is speciation analysis?
Speciation analysis is the process of separating and quantifying different molecular versions of a compound, which can exhibit very different physiochemical properties, including varying toxicities.
Which ICP-OES and ICP-MS instruments are available from Thermo Fisher Scientific?
Thermo Fisher Scientific has over thirty years of experience in designing and manufacturing ICP systems. We offer both ICP-OES and ICP-MS products. Our ICPOES products offer both dedicated radial and dual radial/axial views. In our ICP-MS portfolio, we offer both single and triple quadrupole mass spectrometers together with high-resolution magnetic sector mass spectrometers:
- Thermo Scientific iCAP 7000 Plus ICP-OES Series (https://www.thermofisher.com/uk/en/home/industrial/spectroscopy-elemental-isotope-analysis/trace-elemental-analysis/inductively-coupled-plasma-optical-emission-spectrometry-icp-oes.html)
- Thermo Scientific iCAP RQ ICP-MS (https://www.thermofisher.com/order/catalog/product/IQLAAGGAAQFAQKMBIT)
- Thermo Scientific iCAP TQ ICP-MS (https://www.thermofisher.com/order/catalog/product/731546)
- Thermo Scientific ELEMENT 2 High-Resolution ICP-MS Series (https://www.thermofisher.com/order/catalog/product/IQLAAEGAAMFABWMAFB)
- Thermo Scientific ELEMENT XR High-Resolution ICP-MS (https://www.thermofisher.com/order/catalog/product/IQLAAEGAAMFABWMAFB)
The iCAP RQ ICP-MS (https://www.thermofisher.com/order/catalog/product/IQLAAGGAAQFAQKMBIT) is the recommended model for pharmaceutical elemental impurities.
Is there any instrument that can measure the heavy metals in raw materials or solids?
Laser ablation (LA) is a tool for the direct analysis of solid materials with ICP-MS. There are however some limiting factors to the use of LA-ICP-MS for the direct analysis of solid material, for example the homogeneity of the sample. Furthermore, the sample needs to resist the impact of the laser beam without decomposition.
We have recently purchased an ICP-MS from Thermo Fisher Scientific. Can I connect my existing chromatography systems to it for speciation?
Yes, the interfacing of chromatography to our ICP-MS products is straightforward. In principle, the column ending is connected directly to the nebulizer of the ICP-MS system. Qtegra ISDS software (https://www.thermofisher.com/order/catalog/product/IQLAAEGABSFAOVMBCZ) contains a plug-in for Thermo Scientific Chromeleon Chromatography Data System (CDS) software (https://www.thermofisher.com/order/catalog/product/CHROMELEON7) in order to seamlessly integrate and control Thermo Scientific ion chromatography (IC), ultra high performance liquid chromatography (UHPLC), or gas chromatography (GC) products. In this case, creation of the method and the sample list can be accomplished in one single software window, with automatic triggering of data acquisition. For chromatography systems from other vendors, a trigger cable can be used to start and stop acquisition.
In which instances would a triple quadrupole instrument be required for testing of elemental impurities?
Triple quadrupole ICP-MS instruments use an advanced interference removal mechanism and are capable of achieving LOD for some elements in challenging sample matrices. For typical samples as they are found for elemental impurities analysis, single quadrupole instruments are perfectly capable of removing all typical interferences. However, triple quadrupole ICP-MS offers advantages for the analysis of heavily interfered analytes, such as phosphorous (31P) or sulphur (32S), and can therefore be a valuable tool when used in researc-focused laboratories.
What effects may compromise my analysis results using ICP-OES or ICP-MS?
Interferences are of most concern in ICP-MS. One can basically distinguish two types of interferences: spectral interferences and physical interferences. Mostly, polyatomic interferences are observed, but can mostly be efficiently suppressed by using an instrument equipped with a collision/reaction cell system.
Other interferences observed are isobaric interferences, for example, caused by two elements present in a sample that have isotopes with a common mass number. Mathematical correction using another isotope of the interfering element and calculating the contribution to the signal by means of the isotopic abundance is a way to overcome these interferences.
Doubly charged ions can interfere with some elements, especially if a high concentration of an element with a low 2nd ionization potential is present in a sample, e.g., samarium (Sm), a member of the rare earth elements, that would interfere with the detection of As at m/z 75.
In addition, ionization effects may lead to false positives, for example, if carbon is present in a sample (e.g., after direct dissolution of a sugar containing product in water), As may show an elevated response. Microwave digested samples are normally not affected.
Can I use ICP-OES or ICP-MS as a screening tool?
Both ICP-OES as well as ICP-MS can acquire full spectra to identify all elements present in a sample. This function is not only useful for method development where it helps to identify the origin of interference but it can also be used as part of a risk-based testing approach. If suppliers issue certificates for given elements, these can be removed from quantitative testing, and instead be systematically screened for in a spot-testing approach to assure compliance at all times.
Can I use autodilution to help in the preparation of standards and samples for ICP-OES and ICP-MS analysis?
Yes, autodilution can be used on both ICP-OES and ICP-MS to automate and streamline preparation of calibration standards and perform final sample dilutions. Autodilution systems are also based on valve systems similar to segmented flow introduction systems and are capable of generating different calibration curves (e.g., for drug products with varying daily dosage) from a single stock solution. This reduces the workload for laboratory personnel and also reduces the amount of human interaction with the samples. If a given sample exceeds the calibration range or leads to an internal standard response outside the allowed acceptance criteria, a sample can be automatically diluted and the analysis is repeated. At the same time, autodilution systems are completely integrated into the Qtegra ISDS (https://www.thermofisher.com/order/catalog/product/IQLAAEGABSFAOVMBCZ) operating software, so that all dilution (prescriptive or automatic) steps are documented in a compliant software environment.
What is a segmented flow sample introduction system and how can it help me?
Segmented flow sample introduction systems are a very helpful tool to increase the productivity in your laboratory. Such devices work with a valve system that enables fast uptake of the sample to the plasma, and minimize the wash out times between different samples. Therefore, the time required for analysis can be shortened considerably. An example for such a system is the sprint valve system available on the Thermo Scientific iCAP 7600 ICP-OES (https://www.thermofisher.com/order/catalog/product/842320076121).
Can slurry nebulization be used with pharmaceutical products?
Slurry nebulization is more commonly used in environmental testing applications. In principle, slurry nebulization can be used for pharmaceutical testing and there have been studies done on this. Obviously particle size is a key factor here - the smaller the particles the better. However, it is also worth pointing out that the method preferred by USP is microwave digestion.
Is it also possible to run samples prepared in organic solvents on an ICP-MS or is it just an option for ICP-OES?
It is possible to run samples prepared in organic solvents using ICP-MS. In contrast to ICP-OES, some additional measures have to be taken to achieve optimum performance of the instrument. In particular, the ICP-MS instrument must be fitted with an organics kit which includes a mass flow controller for introducing an Ar/oxygen (O2) mixture into the plasma, to burn up the carbon in the solvent to prevent it from condensing on the cones.
How much more difficult is it to use ICP-MS in comparison to ICP-OES?
Modern ICP-MS instrumentation has become far easier to operate, both in terms of hardware and software. Furthermore, the advent of intelligent interference removal features such as “low mass cut-off” and kinetic energy discrimination (KED) mode has more or less overcome the need to worry about which settings (cell gas, operation mode, cell operating parameters) are appropriate for which element. Overall, the barrier to success with ICP-MS has been lowered significantly and it should not be viewed as a technically challenging alternative to ICP-OES.
When analyzing elemental impurities in phramaceutical samples, how is the method detection limit calculated and how is it related to the parameter instrumental detection limit and blank equivalent concentration?
The parameters BEC, LOD, and LOQ are frequently used to describe the detection capabilities of an analytical instrument. The acronym BEC abbreviates the parameter blank equivalent concentration. This value refers to the “apparent concentration”, and is composed of the contamination level in the blank, any residual interference signal, and the instrument background (from the detection system). The parameter instrumental detection limit (IDL, often referred to as Limit of Detection, LOD) is defined as the limit of detection that can be achieved by the instrument used. This amount is typically defined as a quantity that gives a distinguishable signal in the detection system. The common definition for the IDL is based upon the standard deviation (LOD = 3.3 x standard deviation of the regression line of calibration curve) of a blank sample measured in the beginning of a calibration curve, or a minimum signal to noise ratio of 3:1. Both parameters, BEC and IDL are automatically calculated by Thermo Scientific Qtegra Intelligent Scientific Data Solution (ISDS) platform software (https://www.thermofisher.com/order/catalog/product/IQLAAEGABSFAOVMBCZ). In contrast, on the IDL, the parameter method detection limit (MDL) includes the extent of all dilution steps carried out during the sample preparation. The MDL is typically based upon a blank solution that has been prepared according to the preparation procedure that is being used to prepare all the samples. In situations where there is no sample preparation or if the preparation involves a single dilution step, MDLs might be calculated based on the standard deviation of a low level standard. MDL concentrations are always more conservative than IDL concentrations.
How does the format of my sample impact dilution when analyzing elemental impurities in pharmaceutical drug products?
If samples are in solid form, the dilution incurred will be dictated by the digestion procedure being used to prepare the sample for analysis. If samples are being analyzed in their native form or after simple dilution, there may be more flexibility in the dilution factor used during preparation. Regardless of the sample's original form, the sample matrix must contain a tolerable level of dissolved solids prior to introducing it into the instrument. If an ICP-MS is being used for analysis, the sample matrix should contain 0.2% TDS or, if no special configuration for the sample introduction system is used (e.g., utilizing AGD).
How much should I dilute my samples before analysis using ICP-MS?
ICP-MS is a highly sensitive technique for the detection of elemental impurities. The required limits set out by USP chapter <232> are far higher than the levels that can be detected with an ICP-MS, so dilution can be accomplished easily without compromising performance. You should make sure that samples are diluted such that all analytes are at concentrations well within the calibrated concentration ranges. It should also be noted that analytes shouldn't be diluted such that their concentrations are too close to the instrument's detection limit. It is important to remember that maintaining a clean instrument is a mandatory prerequisite to long-term performance of the machine.
Hg, for example, is well known to show memory effects that increase wash out times or even lead to overestimation of the content of subsequent samples. It is therefore recommended to start with a higher dilution factor during method development and decrease if required.
What is the best choice for an internal standard to be used with pharmaceutical samples?
Internal standards are used in many applications to correct for potential drifts in instrumental sensitivity over time or changes in the sample matrix. The selection of a suitable internal standard should include the following aspects: - The internal standard should have a first ionization potential similar to the analyte, and should have a similar mass as the analyte.
- The internal standard must not be part of the sample.
- It should not generate or be affected by spectral interferences.
- It should be at a low and uniform (preferably zero) concentration in all samples.
Although ICP-MS is a technique considered by some to be relatively robust with respect to matrix effects, in reality, matrix effects do commonly exist and the use of internal standards is standard practice. Internal standards also help account for changes in the transport efficiency of the sample through aerosol.
What is the required acid purity that I should use in my laboratory for pharmaceutical drug products?
The purity of the acids and reagents used is a crucial factor for the attainable method's performance. For ICP-OES, certified trace metal grade is usually sufficient whereas ultra-trace grade is required to leverage the full potential of your ICP-MS system. Also the purity of the water used to prepare diluents and dilutions is a limiting factor. If water (or any solution used in elemental impurities testing) should be stored for some time, Teflon containers should be used in order to avoid contamination of the solutions. All glassware used for sample preparation should be well rinsed with water or dilute acid and the water/dilute acid should be of high purity so as not to introduce contaminants while cleaning the glassware.
Which acid is best to prepare pharmaceutical samples?
The choice of acid is dependent upon the constituent of the drug e.g., if a silicone excipient is present, hydrofluoric acid (HF) will be required and special safety precautions should be followed*. Many people believe that nitric acid (HNO3) is a universal solution which is not entirely correct. Nonetheless, nitric acid (maybe in combination with hydrogen peroxide) is a good acid to start with when organic material needs to be digested. Sulphuric acid (H2SO4) can be used as a means to increase reaction temperature relative to pressure. Addition of hydrochloric acid (HCl) is often used to stabilize Hg and Pt group elements (PGEs). For more help with choosing an appropriate acid, consult the manufacturer of your closed vessel digestion system as they usually have a “cookbook” available. Key to the use of the cookbook is knowing what is contained in the drug i.e., get a full listing of the excipients and APIs. Details of reagents for trace elemental analysis can be found here (http://www.thermofishersci.in/lit/Fisher%20Chemical%20High%20Purity%20Acids.pdf). *Special safety precautions must be taken when using HF. Please refer to local guidelines to ensure all safety requirements are met.
Which instrument type should I choose, one based on (ICP-optical emission spectroscopy) or ICP-MS (ICP-mass spectrometry)?
Both techniques are suitable for meeting the United States Pharmacopeial Convention (USP) Chapter <232> concentration limits, but the sample preparation procedure may dilute the analytes of interest to concentrations below the detection limits of an ICP-OES. ICP-MS provides a larger dynamic range, which may be a requirement for certain applications.
What is the difference between ICP-OES (ICP-optical emission spectroscopy) and ICP-MS (ICP-mass spectrometry)?
Both techniques have a similar excitation source for the sample, an inductively coupled plasma. In ICP-OES, the atoms and ions in the sample are excited by the energy provided by the plasma. After relaxation to the ground state, the atoms emit light of a characteristic wavelength, which is then separated in an optical system and detected by a photosensitive chip.
In ICP-MS, the sample is ionized and the ions are separated according to their mass/charge (m/z) ratio. Also here, each element gives a characteristic set of signals (the isotopic fingerprint). For both ICP-OES and ICP-MS, the signal intensity for each element is directly proportional to its concentration in the measured sample.
What is an inductively coupled plasma (ICP)?
In addition to solid, liquid, and gas, plasma is sometimes considered as a fourth state of matter. A plasma is formed when a gas contains a significant amount of ions and free electrons. Plasmas can be high energy, high temperature entities with temperatures of up to 10,000 degrees C. An example of a high energy plasma is a lightning flash.
In an ICP as used in ICP-optical emission spectroscopy (OES) and ICP-mass spectrometry (MS) instruments, argon (Ar) is used to generate the plasma inside a torch assembly made from quartz. The required energy (typically 1500 W) is provided through an RF frequency and applied via an induction coil surrounding the torch. The plasma is ignited using a short electrical discharge and maintained as long as the RF frequency is applied. The sample is introduced through the central channel of the torch and hence directly entering the plasma. All parts of the system are either water-cooled, or cooled using Ar at a higher flow rate.
What is an elemental impurity?
Elemental impurities are traces of metals that can be found in finished drug products. The analysis of elemental impurities is necessary to describe the concentration of the trace elements in final drug products. Some elements, such as arsenic (As) or lead (Pb), may be present in a drug product based on their natural occurrence. Others, such as palladium (Pd) or platinum (Pt) may be present as residues from catalysts which are used in active pharmaceutical ingredient (API) manufacture. Some elements are intentionally added to a drug product, whereas others leach into the drug product from the drug packaging, or, in the case of elements such as nickel (Ni) or copper (Cu), stem from manufacturing equipment. Due to their potentially adverse effects in the human body, and unknown effects on the stability and effectiveness of a drug product, the concentration of elemental impurities has to be determined in all drug products.