Search
View additional product information for NanoDrop™ Reagents - FAQs (CHEM-PV-8, CHEM-CF-8, CHEM-PR1-KIT, CHEM-PV-1, CHEM-CF-1)
1 product FAQs found
Yes. While most users will never need to recalibrate their NanoDrop One/Onec Spectrophotometer, we recommend verifying instrument performance using the NanoDrop PV-1 Performance Verification Solution every 6 months. PV-1 is available from the Thermo Fisher website.