Sponge Pad for Blotting - FAQs
View additional product information for Sponge Pad for Blotting - FAQs (EI9052)
5 product FAQs found
What are the dimensions of the sponge pads you offer for blotting?
The dimensions of the sponge pads are 8 cm x 9 cm.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Do you offer replacement parts for the XCell II Blot Module?
Except for the sponge pads (Cat. No. EI9052), none of the other parts are available separately.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Should I clean the sponge pads after each use and how should I do it?
We recommend cleaning the sponge pads after each use by rinsing with deionized water and squeezing the water out of the sponge pad 3-5 times.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I purchase replacement parts for the XCell SureLock Mini-Cell?
Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the components of the XCell SureLock Mini-Cell?
The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.