Our facility does not allow flow cytometry of live cells. Can I fix the cells after staining with MitoTracker Deep Red FM dye?
Yes, this is possible because MitoTracker Deep Red FM dye is retained upon fixation. See Table 2 in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/mp07510.pdf).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.