Search
View additional product information for Nuclear Yellow (Hoechst S769121, trihydrochloride, trihydrate) - FAQs (N21485)
1 product FAQs found
DAPI is a very common blue-fluorescent dye for nuclear counterstaining and gives very bright labeling on nuclei in fixed and permeabilized cells and tissues. However, it is considered to be a semi-permeant to impermeant stain and provides inconsistent staining of live cells. Hoechst 33342 dye is cell-permeant and stains with the same binding mechanism and fluorescent color; it is preferred for live-cell imaging and is just as good as DAPI for fixed cell labeling.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.