YO-PRO™-3 Iodide (612/631), 1 mM Solution in DMSO, 1 mL - FAQs
View additional product information for Monomeric Cyanine Nucleic Acid Stains - FAQs (Y3603, P3581, T3605, Y3607, T3602)
2 product FAQs found
What is the suggested working concentration for Monomeric Cyanine Nucleic Acid Stains (Cat. Nos. T3605, P3581, T3602, Y3603, Y3607) for fluorescence microscopy?
For fluorescence microscopy, the working concentration of Monomeric Cyanine Nucleic Acid Stains (Cat. Nos. T3605, P3581, T3602, Y3603, Y3607) is typically 1 - 10 µM. You may need to titrate the dye solution to optimize the working concentration for your sample type and application.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the suggested working concentration for Monomeric Cyanine Nucleic Acid Stains (Cat. Nos. T3605, P3581, T3602, Y3603, Y3607) for flow cytometry?
For flow cytometry, the working concentration of Monomeric Cyanine Nucleic Acid Stains (Cat. Nos. T3605, P3581, T3602, Y3603, Y3607) is typically 25 nM - 1 µM. You may need to titrate the dye solution to optimize the working concentration for your sample type and application.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.