Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101723.4 | NP_564059.1 | 3 - 4 | 1099 | 66 | |
GenBank mRNA | AK228238.1 | - | 3 - 4 | 434 | 66 | |
AY087143.1 | - | 3 - 4 | 441 | 66 | ||
BT006195.1 | - | 3 - 4 | 324 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |